Product Details

SNP ID
rs151287767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:617466 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTCCTTCAGCTGGCTGGCCTCC[A/G]AGATCTTCTGGGGCCTGGGGTTGGA
Phenotype
MIM: 602781 MIM: 601778
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HCN2 PubMed Links

Gene Details

Gene
HCN2
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001194.3 Intron NP_001185.3
Gene
POLRMT
Gene Name
RNA polymerase mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005035.3 Intron NP_005026.3
XM_005259580.3 Intron XP_005259637.2

View Full Product Details