Product Details

SNP ID
rs111337477
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:217643187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCATTCTAGTTTTTTTCTCTTAC[A/C]TTTTCTGACAGAGAGAAACCTGGCT
Phenotype
MIM: 611032
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SPATA17 PubMed Links

Gene Details

Gene
SPATA17
Gene Name
spermatogenesis associated 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138796.3 Intron NP_620151.1
XM_005273052.1 Intron XP_005273109.1
XM_006711165.3 Intron XP_006711228.1
XM_011509194.2 Intron XP_011507496.1

View Full Product Details