Product Details
- SNP ID
-
rs111438636
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
13
- Location
-
Chr.1:42736197 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGTGAATGCTCTTCCTTTCTCCCT[A/C]CCCCCCAGCCCAGAATATCAACACC
- Phenotype
-
MIM: 610036
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CLDN19
PubMed Links
Gene Details
- Gene
- CLDN19
- Gene Name
- claudin 19
View Full Product Details