Product Details

SNP ID
rs111476958
Assay Type
Functionally Tested
NCBI dbSNP Submissions
14
Location
Chr.1:241532458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCAGCAGTTATGGACTCATCTG[C/T]CATTCAAAGGAAAAAAGTAGCTGTC
Phenotype
MIM: 603538
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KMO PubMed Links

Gene Details

Gene
KMO
Gene Name
kynurenine 3-monooxygenase (kynurenine 3-hydroxylase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003679.4 435 Missense Mutation GCC,GTC A5V NP_003670.2
XM_005273337.1 435 Missense Mutation GCC,GTC A45V XP_005273394.1
XM_005273338.3 435 UTR 5 XP_005273395.1
XM_017002649.1 435 UTR 5 XP_016858138.1
XM_017002650.1 435 Intron XP_016858139.1

View Full Product Details