Product Details

SNP ID
rs111589211
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:46344804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCTGTCCTCTCTCTTTTAGGCT[A/G]CCACAGAGCCCAAATTCCCTGCTGT
Phenotype
MIM: 615394
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NSUN4 PubMed Links

Gene Details

Gene
NSUN4
Gene Name
NOP2/Sun RNA methyltransferase family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256127.1 324 UTR 5 NP_001243056.1
NM_001256128.1 324 UTR 5 NP_001243057.1
NM_199044.3 324 Missense Mutation ACC,GCC T33A NP_950245.2

View Full Product Details