Product Details

SNP ID
rs111673139
Assay Type
Functionally Tested
NCBI dbSNP Submissions
14
Location
Chr.1:182056043 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTCCCCAACCCAAGGAACTCAA[C/T]GTTTGATCAGCTCCCACTACTTTGT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZNF648 PubMed Links

Gene Details

Gene
ZNF648
Gene Name
zinc finger protein 648
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009992.1 6109 UTR 3 NP_001009992.1
XM_017000301.1 6109 UTR 3 XP_016855790.1
XM_017000302.1 6109 UTR 3 XP_016855791.1

View Full Product Details