Product Details

SNP ID
rs111746408
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:66926216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTAAGAACCAACTGGTTGAGGTTC[A/C]ATGCAGACAAGACGGATGTGATGCT
Phenotype
MIM: 616848
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MIER1 PubMed Links

Gene Details

Gene
MIER1
Gene Name
MIER1 transcriptional regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077700.2 271 Missense Mutation AAT,CAT N48H NP_001071168.2
NM_001077701.2 271 Intron NP_001071169.1
NM_001077702.2 271 UTR 5 NP_001071170.2
NM_001077703.2 271 Missense Mutation AAT,CAT N48H NP_001071171.2
NM_001077704.2 271 Intron NP_001071172.1
NM_001146110.1 271 UTR 5 NP_001139582.1
NM_001146111.1 271 UTR 5 NP_001139583.1
NM_001146112.1 271 UTR 5 NP_001139584.1
NM_001146113.1 271 UTR 5 NP_001139585.1
NM_001278215.1 271 UTR 5 NP_001265144.1
NM_020948.3 271 UTR 5 NP_065999.2
XM_005271076.3 271 UTR 5 XP_005271133.1
XM_011541864.2 271 Missense Mutation AAT,CAT N48H XP_011540166.1
XM_011541865.2 271 Missense Mutation AAT,CAT N48H XP_011540167.1
XM_017001919.1 271 UTR 5 XP_016857408.1
XM_017001920.1 271 UTR 5 XP_016857409.1
XM_017001921.1 271 UTR 5 XP_016857410.1
XM_017001922.1 271 UTR 5 XP_016857411.1
XM_017001923.1 271 Missense Mutation AAT,CAT N48H XP_016857412.1
XM_017001924.1 271 UTR 5 XP_016857413.1
XM_017001925.1 271 UTR 5 XP_016857414.1
XM_017001926.1 271 UTR 5 XP_016857415.1
XM_017001927.1 271 UTR 5 XP_016857416.1
XM_017001928.1 271 UTR 5 XP_016857417.1
XM_017001929.1 271 UTR 5 XP_016857418.1
XM_017001930.1 271 Intron XP_016857419.1
XM_017001931.1 271 UTR 5 XP_016857420.1
XM_017001932.1 271 UTR 5 XP_016857421.1
XM_017001933.1 271 Intron XP_016857422.1
XM_017001934.1 271 Intron XP_016857423.1
Gene
WDR78
Gene Name
WD repeat domain 78
There are no transcripts associated with this gene.

View Full Product Details