Product Details

SNP ID
rs112073592
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:27347472 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGCAGCTGAAGATCCTGACAGG[A/G]GACTGGTTCCAGGAAGCACGCTCCC
Phenotype
MIM: 604468 MIM: 608042
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAP3K6 PubMed Links

Gene Details

Gene
MAP3K6
Gene Name
mitogen-activated protein kinase kinase kinase 6
There are no transcripts associated with this gene.

Gene
SYTL1
Gene Name
synaptotagmin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193308.1 440 Silent Mutation GGA,GGG G81G NP_001180237.1
NM_032872.2 440 Silent Mutation GGA,GGG G81G NP_116261.1
XM_005246022.3 440 Silent Mutation GGA,GGG G113G XP_005246079.1
XM_006710990.1 440 Silent Mutation GGA,GGG G113G XP_006711053.1
XM_006710991.1 440 UTR 5 XP_006711054.1

View Full Product Details