Product Details

SNP ID
rs112421279
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:39659132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCGGTGGTTCAGCTACTGTGCAG[A/C]TGGGGCCTGCTTTGCAGGTGCAGTC
Phenotype
MIM: 610525
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NT5C1A PubMed Links

Gene Details

Gene
NT5C1A
Gene Name
5'-nucleotidase, cytosolic IA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032526.2 1096 Missense Mutation GCT,TCT A366S NP_115915.1

View Full Product Details