Product Details

SNP ID
rs112422735
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:231475975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGGGAGGTGGCCCCCACTTGTT[A/G]GTGATGACTGAGTCCCTGCACAGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNRPD2P2 PubMed Links

Gene Details

Gene
SNRPD2P2
Gene Name
small nuclear ribonucleoprotein D2 pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details