Product Details

SNP ID
rs112429287
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:116383617 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTGAGAATGCGGACTCATAAGC[A/G]CAATATTGCACTCTAAGTTAATAAT
Phenotype
MIM: 182310
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP1A1 PubMed Links

Gene Details

Gene
ATP1A1
Gene Name
ATPase Na+/K+ transporting subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000701.7 Intron NP_000692.2
NM_001160233.1 Intron NP_001153705.1
NM_001160234.1 Intron NP_001153706.1
XM_017001360.1 Intron XP_016856849.1
XM_017001361.1 Intron XP_016856850.1
Gene
ATP1A1-AS1
Gene Name
ATP1A1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details