Product Details

SNP ID
rs112527476
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:231476130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGCACCATGTTGCAGTGTCTAC[C/T]CAAGGCCTTCGCAGCCCAGCAGCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNRPD2P2 PubMed Links

Gene Details

Gene
SNRPD2P2
Gene Name
small nuclear ribonucleoprotein D2 pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details