Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004388.2 | 1418 | UTR 3 | NP_004379.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001310156.1 | 1418 | Intron | NP_001297085.1 | ||
XM_011542514.2 | 1418 | Intron | XP_011540816.1 | ||
XM_011542515.2 | 1418 | Intron | XP_011540817.1 | ||
XM_011542516.1 | 1418 | Intron | XP_011540818.1 | ||
XM_011542517.1 | 1418 | Intron | XP_011540819.1 | ||
XM_011542519.2 | 1418 | Intron | XP_011540821.1 | ||
XM_011542520.2 | 1418 | Intron | XP_011540822.1 | ||
XM_011542521.2 | 1418 | Intron | XP_011540823.1 | ||
XM_011542522.2 | 1418 | Intron | XP_011540824.1 |