Product Details

SNP ID
rs112656652
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:42927244 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACGTAGGGACCACACAGTTGCT[C/G]AAAGACACACAGACACACTTGGTTG
Phenotype
MIM: 138140 MIM: 143090
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC2A1 PubMed Links

Gene Details

Gene
SLC2A1
Gene Name
solute carrier family 2 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006516.2 Intron NP_006507.2

View Full Product Details