Product Details

SNP ID
rs112666836
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:151518765 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGTGACTCCTTTGGGGTCCAAAT[C/T]AAGGGGGCCAATGACCAAGGGGCCT
Phenotype
MIM: 609473
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CGN PubMed Links

Gene Details

Gene
CGN
Gene Name
cingulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020770.2 390 Silent Mutation ATC,ATT I82I NP_065821.1
XM_005245365.4 390 Missense Mutation ATC,ATT I82I XP_005245422.1

View Full Product Details