Product Details

SNP ID
rs112752846
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:47218451 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGTGATTTACAAAAATTTCATG[C/T]ACAACTTTCAGAGGGGAGCACTTCT
Phenotype
MIM: 187040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAL1 PubMed Links

Gene Details

Gene
TAL1
Gene Name
TAL bHLH transcription factor 1, erythroid differentiation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287347.2 2591 UTR 3 NP_001274276.1
NM_001290403.1 2591 UTR 3 NP_001277332.1
NM_001290404.1 2591 UTR 3 NP_001277333.1
NM_001290405.1 2591 UTR 3 NP_001277334.1
NM_001290406.1 2591 UTR 3 NP_001277335.1
NM_003189.5 2591 UTR 3 NP_003180.1
XM_005271160.4 2591 UTR 3 XP_005271217.1
XM_017002187.1 2591 UTR 3 XP_016857676.1
XM_017002188.1 2591 UTR 3 XP_016857677.1
XM_017002189.1 2591 UTR 3 XP_016857678.1
XM_017002190.1 2591 UTR 3 XP_016857679.1
XM_017002191.1 2591 UTR 3 XP_016857680.1
XM_017002192.1 2591 Intron XP_016857681.1
XM_017002193.1 2591 UTR 3 XP_016857682.1

View Full Product Details