Product Details

SNP ID
rs112753945
Assay Type
Functionally Tested
NCBI dbSNP Submissions
13
Location
Chr.1:63368924 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAACCACTACTCAAATCAAGAAG[C/G]AAGATTCACAGCATTTCAGAACACC
Phenotype
MIM: 604566
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ALG6 PubMed Links

Gene Details

Gene
ALG6
Gene Name
ALG6, alpha-1,3-glucosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013339.3 Intron NP_037471.2

View Full Product Details