Product Details

SNP ID
rs112826258
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:156285318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAGGCCTTGGTTCCCAATGGCC[A/G]GCAGCCAGAAGGGGAAGGTGGGGCC
Phenotype
MIM: 610962 MIM: 615531
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLMP PubMed Links

Gene Details

Gene
GLMP
Gene Name
glycosylated lysosomal membrane protein
There are no transcripts associated with this gene.

Gene
SMG5
Gene Name
SMG5, nonsense mediated mRNA decay factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323614.1 263 Intron NP_001310543.1
NM_001323615.1 263 Intron NP_001310544.1
NM_001323616.1 263 Intron NP_001310545.1
NM_001323617.1 263 Intron NP_001310546.1
NM_015327.2 263 Intron NP_056142.2
XM_017000842.1 263 Intron XP_016856331.1
XM_017000843.1 263 Intron XP_016856332.1
Gene
TMEM79
Gene Name
transmembrane protein 79
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032323.2 263 Missense Mutation CAG,CGG Q31R NP_115699.1

View Full Product Details