Product Details

SNP ID
rs112941181
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:248362224 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCATTTCTTCTGTGAAGTTCCTG[C/G]TGTATTGAATCTCTCCTGCTCAGAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR2T4 PubMed Links

Gene Details

Gene
OR2T4
Gene Name
olfactory receptor family 2 subfamily T member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004696.1 Intron NP_001004696.1

View Full Product Details