Product Details

SNP ID
rs112946539
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:39770456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATTTCTGCCCCTTTATCACGCGA[C/T]CTACATAAATGTTAGGAACGTGGAT
Phenotype
MIM: 602284 MIM: 610289 MIM: 602435
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BMP8B PubMed Links

Gene Details

Gene
BMP8B
Gene Name
bone morphogenetic protein 8b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005271149.4 Intron XP_005271206.1
XM_011542022.2 Intron XP_011540324.1
XM_011542024.2 Intron XP_011540326.2
XM_011542025.2 Intron XP_011540327.1
XM_017002155.1 Intron XP_016857644.1
XM_017002156.1 Intron XP_016857645.1
Gene
OXCT2
Gene Name
3-oxoacid CoA-transferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022120.1 Intron NP_071403.1
Gene
PPIE
Gene Name
peptidylprolyl isomerase E
There are no transcripts associated with this gene.

View Full Product Details