Product Details

SNP ID
rs113005319
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:1336473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCCGACGCTCCTTCTCACGGCCC[C/G]GGGCCCGGCGGCTGCTCCGGGTGGA
Phenotype
MIM: 615467 MIM: 601365 MIM: 605865
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CPTP PubMed Links

Gene Details

Gene
CPTP
Gene Name
ceramide-1-phosphate transfer protein
There are no transcripts associated with this gene.

Gene
DVL1
Gene Name
dishevelled segment polarity protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004421.2 1834 Missense Mutation CCG,CGG P561R NP_004412.2
XM_005244731.3 1834 Missense Mutation CCG,CGG P586R XP_005244788.1
XM_005244732.3 1834 UTR 3 XP_005244789.1
XM_005244733.3 1834 UTR 3 XP_005244790.1
Gene
MIR6808
Gene Name
microRNA 6808
There are no transcripts associated with this gene.

Gene
TAS1R3
Gene Name
taste 1 receptor member 3
There are no transcripts associated with this gene.

View Full Product Details