Product Details

SNP ID
rs113071219
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:225937464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTATGGCTGGAGCCTCCTTGGCA[C/T]GAGCGCCCCATCCGAGGCACAGCTG
Phenotype
MIM: 601877
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LEFTY2 PubMed Links

Gene Details

Gene
LEFTY2
Gene Name
left-right determination factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011544266.1 Intron XP_011542568.1

View Full Product Details