Product Details

SNP ID
rs113101028
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:16977161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACGTGGTCAGGGAACGGCGGCAG[C/T]GGGTCCAGGTCATACTGGCCCTGAG
Phenotype
MIM: 610513 MIM: 615776 MIM: 156790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP13A2 PubMed Links

Gene Details

Gene
ATP13A2
Gene Name
ATPase 13A2
There are no transcripts associated with this gene.

Gene
CROCC
Gene Name
ciliary rootlet coiled-coil, rootletin
There are no transcripts associated with this gene.

Gene
MFAP2
Gene Name
microfibrillar associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135247.1 186 Silent Mutation CCA,CCG P24P NP_001128719.1
NM_001135248.1 186 Silent Mutation CCA,CCG P24P NP_001128720.1
NM_002403.3 186 Silent Mutation CCA,CCG P25P NP_002394.1
NM_017459.2 186 Silent Mutation CCA,CCG P25P NP_059453.1
XM_011541476.2 186 Silent Mutation CCA,CCG P25P XP_011539778.1

View Full Product Details