Product Details

SNP ID
rs113101902
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:25245162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGAATACTTACTTCAGGCTTTGC[A/G]CCATTACTTGATACACCTATTCCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RSRP1 PubMed Links

Gene Details

Gene
RSRP1
Gene Name
arginine and serine rich protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321772.1 902 Silent Mutation GGC,GGT G220G NP_001308701.1
NM_020317.4 902 Silent Mutation GGC,GGT G220G NP_064713.3
XM_011541797.1 902 Silent Mutation GGC,GGT G220G XP_011540099.1

View Full Product Details