Product Details

SNP ID
rs113408337
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:23779458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGTTTGTTGAAAGTGATGCAGA[C/T]GAAGAGCTTCTGTTTAATATTCCGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PITHD1 PubMed Links

Gene Details

Gene
PITHD1
Gene Name
PITH domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020362.4 349 Silent Mutation GAC,GAT D73D NP_065095.2
XM_011541804.1 349 Intron XP_011540106.1
Gene
TCEB3-AS1
Gene Name
TCEB3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details