Product Details

SNP ID
rs113457355
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:111414484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCTTAGGCTTCTTCATCCACAG[C/G]AGAGTTTTCTGGGATTTCTTTTTTT
Phenotype
MIM: 603578
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OVGP1 PubMed Links

Gene Details

Gene
OVGP1
Gene Name
oviductal glycoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002557.3 2073 Missense Mutation CCT,GCT P673A NP_002548.3

View Full Product Details