Product Details

SNP ID
rs113552832
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:22658382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCCTAATCATCACTTCCAAGAG[G/T]GGCTTCCCCTTGGAGAAGACTTTCC
Phenotype
MIM: 120570
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C1QB PubMed Links

Gene Details

Gene
C1QB
Gene Name
complement C1q B chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000491.3 Intron NP_000482.3
XM_011542059.2 Intron XP_011540361.1

View Full Product Details