Product Details

SNP ID
rs113620024
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:150966475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACACAATGGGAACAGGGCTTCA[C/G]CTTTCCAGTTATGAACTTGTGGGCC
Phenotype
MIM: 606920 MIM: 604396
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CERS2 PubMed Links

Gene Details

Gene
CERS2
Gene Name
ceramide synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022075.4 Intron NP_071358.1
NM_181746.3 Intron NP_859530.1
XM_011509451.2 Intron XP_011507753.1
XM_011509452.2 Intron XP_011507754.1
Gene
SETDB1
Gene Name
SET domain bifurcated 1
There are no transcripts associated with this gene.

View Full Product Details