Product Details

SNP ID
rs113738580
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:205659591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTAGGGAAGGGAGCTCCAGGCTT[A/C]GGGCCTGGCAGGAAGCTGGTCATCA
Phenotype
MIM: 605097
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC45A3 PubMed Links

Gene Details

Gene
SLC45A3
Gene Name
solute carrier family 45 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033102.2 1810 Silent Mutation CCG,CCT P435P NP_149093.1
XM_005245556.3 1810 Silent Mutation CCG,CCT P435P XP_005245613.1
XM_005245557.4 1810 Silent Mutation CCG,CCT P435P XP_005245614.1
XM_005245560.2 1810 Intron XP_005245617.1
XM_017002638.1 1810 Silent Mutation CCG,CCT P435P XP_016858127.1

View Full Product Details