Product Details

SNP ID
rs113762641
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:247895743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCATCCTGGTGTCCCGGCTGGAC[A/C]CCCACCTCCACACCCCCATGTACTT
Phenotype
MIM: 616729
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR2W3 PubMed Links

Gene Details

Gene
OR2W3
Gene Name
olfactory receptor family 2 subfamily W member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001957.2 157 Missense Mutation ACC,CCC T53P NP_001001957.2

View Full Product Details