Product Details

SNP ID
rs113946450
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:218285330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCGCAGAAAAATGGCAGCCGCCG[C/T]TCCGGACTCACGTGTGAGTGAGGAA
Phenotype
MIM: 611193
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RRP15 PubMed Links

Gene Details

Gene
RRP15
Gene Name
ribosomal RNA processing 15 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016052.3 44 Missense Mutation GCT,GTT A5V NP_057136.2
XM_011509597.2 44 Missense Mutation GCT,GTT A5V XP_011507899.1

View Full Product Details