Product Details

SNP ID
rs113951378
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:38018019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGGAGGTGCTACATTGAACGTA[C/T]GCCTCAGGGGCCTGTGTCTAGGATT
Phenotype
MIM: 609440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UTP11 PubMed Links

Gene Details

Gene
UTP11
Gene Name
UTP11, small subunit processome component homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016037.3 Intron NP_057121.2
XM_011541555.2 Intron XP_011539857.1

View Full Product Details