Product Details

SNP ID
rs114050025
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:117111259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCAAAGCTCCCAGTTAACTTTC[C/T]AGGTATATAGATAACAGTGAAGACT
Phenotype
MIM: 609318 MIM: 604718
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIM45 PubMed Links

Gene Details

Gene
TRIM45
Gene Name
tripartite motif containing 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145635.1 3377 UTR 3 NP_001139107.1
NM_025188.3 3377 UTR 3 NP_079464.2
XM_011542199.2 3377 UTR 3 XP_011540501.1
Gene
TTF2
Gene Name
transcription termination factor 2
There are no transcripts associated with this gene.

View Full Product Details