Product Details

SNP ID
rs114057591
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:225812747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGGATCACAGCTGGGTGTACCA[A/G]ATAAGGGAGGACCAAGCATGAGCTG
Phenotype
MIM: 132810
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPHX1 PubMed Links

Gene Details

Gene
EPHX1
Gene Name
epoxide hydrolase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000120.3 Intron NP_000111.1
NM_001136018.3 Intron NP_001129490.1
NM_001291163.1 Intron NP_001278092.1

View Full Product Details