Product Details

SNP ID
rs114062490
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:43270453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTGAAGCTGGAGCTAAAGCTAA[A/G]TCGGGGTCGGAGCTGTGGAATGGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM125 PubMed Links

Gene Details

Gene
TMEM125
Gene Name
transmembrane protein 125
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320244.1 Intron NP_001307173.1
NM_144626.2 Intron NP_653227.1
XM_005270467.3 Intron XP_005270524.1
XM_011540705.2 Intron XP_011539007.1

View Full Product Details