Product Details

SNP ID
rs114258996
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:3457604 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAGGAGGGCGGGGCTACAGAGC[A/G]GCCCACCCAACCAACACTGGGTCCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF16 PubMed Links

Gene Details

Gene
ARHGEF16
Gene Name
Rho guanine nucleotide exchange factor 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014448.3 Intron NP_055263.2
XM_017001049.1 Intron XP_016856538.1
XM_017001050.1 Intron XP_016856539.1
XM_017001051.1 Intron XP_016856540.1

View Full Product Details