Product Details

SNP ID
rs114340421
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:40258357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCCGTGCTGCTCTTTTCCTGGA[C/T]AGTGTATCTTTGGGAGACCTTCCTA
Phenotype
MIM: 606480
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMCO2 PubMed Links

Gene Details

Gene
TMCO2
Gene Name
transmembrane and coiled-coil domains 2
There are no transcripts associated with this gene.

Gene
ZMPSTE24
Gene Name
zinc metallopeptidase STE24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005857.4 308 Missense Mutation ACA,ATA T29I NP_005848.2

View Full Product Details