Product Details

SNP ID
rs114522590
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:153373950 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTTGGAATATTTCATCAATGAC[A/G]GCTTTATCTTTGATATTCTAGGAAA
Phenotype
MIM: 603112
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
S100A12 PubMed Links

Gene Details

Gene
S100A12
Gene Name
S100 calcium binding protein A12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005621.1 224 Silent Mutation GCC,GCT A52A NP_005612.1

View Full Product Details