Product Details

SNP ID
rs114545115
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:108562120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTATAAGGTAGGGGTTCCATTT[C/T]GTAGATAAATAAACTGAGGCTCAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM102B PubMed Links

Gene Details

Gene
FAM102B
Gene Name
family with sequence similarity 102 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010883.2 Intron NP_001010883.2

View Full Product Details