Product Details

SNP ID
rs114586414
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:52945101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATTTTGACCCAGACCTAGTCAC[C/T]ATTAGTATCTTGGTGTTATTCTATT
Phenotype
MIM: 184755
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR1273G PubMed Links

Gene Details

Gene
MIR1273G
Gene Name
microRNA 1273g
There are no transcripts associated with this gene.

Gene
SCP2
Gene Name
sterol carrier protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007098.2 Intron NP_001007099.1
NM_001007099.2 Intron NP_001007100.1
NM_001007100.2 Intron NP_001007101.1
NM_001007250.2 Intron NP_001007251.1
NM_001193599.1 Intron NP_001180528.1
NM_001193600.1 Intron NP_001180529.1
NM_001193617.1 Intron NP_001180546.1
NM_002979.4 Intron NP_002970.2
XM_005271103.4 Intron XP_005271160.1
XM_011541935.2 Intron XP_011540237.1
XM_017002045.1 Intron XP_016857534.1
XM_017002046.1 Intron XP_016857535.1

View Full Product Details