Product Details

SNP ID
rs114693585
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:156700551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGGCAGGACTTACCAGGATCAG[C/T]TCCCCATCGTTGGTCAGTTCTCTGG
Phenotype
MIM: 180231
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRABP2 PubMed Links

Gene Details

Gene
CRABP2
Gene Name
cellular retinoic acid binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199723.1 578 Silent Mutation GAA,GAG E119E NP_001186652.1
NM_001878.3 578 Silent Mutation GAA,GAG E119E NP_001869.1
XM_017000343.1 578 Silent Mutation GAA,GAG E119E XP_016855832.1

View Full Product Details