Product Details

SNP ID
rs114751507
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:203497050 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACTACGGCTCCAGGGACACCCT[C/T]GTCAAACCCCACGATGACCAGACCT
Phenotype
MIM: 605127 MIM: 601914
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OPTC PubMed Links

Gene Details

Gene
OPTC
Gene Name
opticin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014359.3 421 Missense Mutation TCG,TTG S102L NP_055174.1
XM_011509406.2 421 Missense Mutation TCG,TTG S102L XP_011507708.1
Gene
PRELP
Gene Name
proline and arginine rich end leucine rich repeat protein
There are no transcripts associated with this gene.

View Full Product Details