Product Details

SNP ID
rs114768585
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:162643921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTTCTTGGGTAGTTAAATTGGA[A/G]AAAGAGAGTCTGGCACATTCAGAAT
Phenotype
MIM: 191311
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDR2 PubMed Links

Gene Details

Gene
DDR2
Gene Name
discoidin domain receptor tyrosine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014796.1 Intron NP_001014796.1
NM_006182.2 Intron NP_006173.2
XM_006711344.3 Intron XP_006711407.1
XM_011509586.2 Intron XP_011507888.1
XM_011509587.2 Intron XP_011507889.1
XM_011509588.2 Intron XP_011507890.1

View Full Product Details