Product Details

SNP ID
rs115041192
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:28148444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGTATGGAAGCCCCTGGACATCG[A/G]GGTGTCCTGGCCTGGACAGCTCCAA
Phenotype
MIM: 173393
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTAFR PubMed Links

Gene Details

Gene
PTAFR
Gene Name
platelet activating factor receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000952.4 2913 UTR 3 NP_000943.1
NM_001164721.1 2913 UTR 3 NP_001158193.1
NM_001164722.2 2913 UTR 3 NP_001158194.1
NM_001164723.2 2913 UTR 3 NP_001158195.1

View Full Product Details