Product Details

SNP ID
rs115057664
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:201468630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGCCTTGATGCGGGCGAAGCTGA[A/G]CTCCTTGGGCCGGCCGCCCGTGCCC
Phenotype
MIM: 607054
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHLDA3 PubMed Links

Gene Details

Gene
PHLDA3
Gene Name
pleckstrin homology like domain family A member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012396.4 542 Missense Mutation CTC,TTC L53F NP_036528.1

View Full Product Details