Product Details

SNP ID
rs115160367
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:247416516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGATATACAGAAGGAAACAAAC[A/C]TCCTTAAAGACCAAAGGGCAGCAGA
Phenotype
MIM: 606416
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NLRP3 PubMed Links

Gene Details

Gene
NLRP3
Gene Name
NLR family pyrin domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079821.2 Intron NP_001073289.1
NM_001127461.2 Intron NP_001120933.1
NM_001127462.2 Intron NP_001120934.1
NM_001243133.1 Intron NP_001230062.1
NM_004895.4 Intron NP_004886.3
NM_183395.2 Intron NP_899632.1
XM_011544048.2 Intron XP_011542350.1
XM_011544053.2 Intron XP_011542355.1
XM_011544055.2 Intron XP_011542357.1
XM_017000181.1 Intron XP_016855670.1
XM_017000182.1 Intron XP_016855671.1
XM_017000183.1 Intron XP_016855672.1
XM_017000184.1 Intron XP_016855673.1

View Full Product Details