Product Details

SNP ID
rs115160731
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:248826432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAAAAGTGACTTTCCTCACTCC[A/C]GTGAAGTCGGCGGAACCCTCCACTA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MIR3124 PubMed Links

Gene Details

Gene
MIR3124
Gene Name
microRNA 3124
There are no transcripts associated with this gene.

Gene
SH3BP5L
Gene Name
SH3 binding domain protein 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322462.1 Intron NP_001309391.1
NM_001322463.1 Intron NP_001309392.1
NM_001322464.1 Intron NP_001309393.1
NM_030645.1 Intron NP_085148.1
XM_017002404.1 Intron XP_016857893.1
XM_017002405.1 Intron XP_016857894.1
XM_017002406.1 Intron XP_016857895.1
XM_017002407.1 Intron XP_016857896.1

View Full Product Details