Product Details

SNP ID
rs115170612
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:9546709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTAGGCTCATTTAGGGGGTGTT[C/T]AGTTGCAAGTAATAGAAGATTCAAC
Phenotype
MIM: 610816
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC25A33 PubMed Links

Gene Details

Gene
SLC25A33
Gene Name
solute carrier family 25 member 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032315.2 Intron NP_115691.1
XM_005263503.3 Intron XP_005263560.1
XM_011542296.1 Intron XP_011540598.1

View Full Product Details