Product Details

SNP ID
rs115262679
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:147638214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAAACACTTAAAAACAAATAATC[A/G]CTAATATTGTTTAGCTTGCCTATGT
Phenotype
MIM: 611471
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACP6 PubMed Links

Gene Details

Gene
ACP6
Gene Name
acid phosphatase 6, lysophosphatidic
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323625.1 Intron NP_001310554.1
NM_016361.4 Intron NP_057445.4
XM_011509601.2 Intron XP_011507903.1
XM_017001422.1 Intron XP_016856911.1

View Full Product Details