Product Details

SNP ID
rs115389934
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:171493919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACAGGCTTTACACTATTCTAAGA[A/G]CTTTATACACACTTTGTTAATTAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRRC2C PubMed Links

Gene Details

Gene
PRRC2C
Gene Name
proline rich coiled-coil 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015172.3 Intron NP_055987.2
XM_005245015.2 Intron XP_005245072.1
XM_005245016.4 Intron XP_005245073.1
XM_005245017.2 Intron XP_005245074.1
XM_005245018.2 Intron XP_005245075.1
XM_005245019.2 Intron XP_005245076.1
XM_005245020.2 Intron XP_005245077.1
XM_005245023.2 Intron XP_005245080.1
XM_005245024.2 Intron XP_005245081.1
XM_005245025.2 Intron XP_005245082.1
XM_005245026.2 Intron XP_005245083.1
XM_005245027.2 Intron XP_005245084.1
XM_005245028.2 Intron XP_005245085.1
XM_005245029.2 Intron XP_005245086.1
XM_006711238.3 Intron XP_006711301.1
XM_017000760.1 Intron XP_016856249.1
XM_017000761.1 Intron XP_016856250.1
XM_017000762.1 Intron XP_016856251.1

View Full Product Details